Benzo[cd]indol-2(1H)-one ≥97.0%

Supplier: TCI America

Synonyms: Naphthostyril, Naphtholactam, Benz[cd]indol-2(1H)-one

B3023-25G B3023-5G
TCB3023-25GEA 327.06 CAD
TCB3023-25G TCB3023-5G
Benzo[cd]indol-2(1H)-one ≥97.0%
Benzo[cd]indol-2(1H)-one

CAS Number: 130-00-7
MDL Number: MFCD00009748
Molecular Formula: C11H7NO
Molecular Weight: 169.18
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Melting point (°C): 181


Other Product Names / Synonyms: Naphthostyril

Formula: C₁₁H₇NO
MW: 169.18 g/mol
Melting Pt: 181 °C
MDL Number: MFCD00009748
CAS Number: 130-00-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56c546f0c32e7c78be99d8e6e1d7ba8239cb1bcd&vid=56c546f0c32e7c78be99d8e6e1d7ba8239cb1bcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR