N,N'-Bis(5-hydroxysalicylidene)ethylenediamine ≥98.0%
Supplier: TCI America
Synonyms:
N,N'-Ethylenebis(5-hydroxysalicylideneimine), N,N-Bis(2,5-dihydroxybenzylidene)ethylenediamine, 2,2'-((Ethane-1,2-diylbis(azanylylidene))bis(methanylylidene))bis(benzene-1,4-diol)
CAS Number: 124061-43-4
MDL Number: MFCD04039747
Molecular Formula: C16H16N2O4
Molecular Weight: 300.31
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Powder
Melting point (°C): 250
Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: N,N'-Ethylenebis(5-hydroxysalicylideneimine)
Formula:
C₁₆H₁₆N₂O₄ MW: 300.31 g/mol Melting Pt: 250 °C |
MDL Number:
MFCD04039747 CAS Number: 124061-43-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...