2,2-Bis(allyloxymethyl)-1-butanol ≥85.0% (contains Mono- and Tri-substituted Product)

Supplier: TCI America

Synonyms: Trimethylolpropane Diallyl Ether

B3003-25G B3003-500G
TCB3003-25GEA 47.4 CAD
TCB3003-25G TCB3003-500G
2,2-Bis(allyloxymethyl)-1-butanol ≥85.0% (contains Mono- and Tri-substituted Product)
2,2-Bis(allyloxymethyl)-1-butanol

CAS Number: 682-09-7
MDL Number: MFCD00035791
Molecular Formula: C12H22O3
Molecular Weight: 214.31
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Flash Point (°C): 124
Specific Gravity (20/20): 0.96


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Trimethylolpropane Diallyl Ether

Formula: C₁₂H₂₂O₃
MW: 214.31 g/mol
Boiling Pt: 122 °C
Density: 0.96 (20 °C)
Flash Pt: 124 °C
MDL Number: MFCD00035791
CAS Number: 682-09-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5620f0a18adffd14062b0adf77e580cb981b4117&vid=5620f0a18adffd14062b0adf77e580cb981b4117&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR