Gallein ≥85.0%
Supplier: TCI America
Synonyms:
Alizarin Violet, 3',4',5',6'-Tetrahydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, Mordant Violet 25
A0601-25G
A0601-1G
TCA0601-025GEA
601.98
CAD
TCA0601-025G
TCA0601-001G
Gallein ≥85.0%
Gallein
CAS Number: 2103-64-2
MDL Number: MFCD00041839
Molecular Formula: C20H12O7
Molecular Weight: 364.31
Purity/Analysis Method: >85.0% (HPLC)
Form: Crystal
Lambda max.: 534 nm (pH 6.8 buffer sol.)
Product Categories: Materials Chemistry, Xanthene Dyes (Functional Organic Dyes)
Other Product Names / Synonyms: Alizarin Violet, Mordant Violet 25
Formula:
C₂₀H₁₂O₇ |
MDL Number:
MFCD00041839 CAS Number: 2103-64-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...