N-(tert-Butoxycarbonyl)-D-glutamic acid ≥97.0%

Supplier: TCI America

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)pentanedioic acid, Boc-D-Glu-OH, N-Boc-D-glutamic Acid

B2987-25G B2987-5G
TCB2987-25GEA 246.32 CAD
TCB2987-25G TCB2987-5G
N-(tert-Butoxycarbonyl)-D-glutamic acid ≥97.0%
N-(tert-Butoxycarbonyl)-D-glutamic acid

CAS Number: 34404-28-9
MDL Number: MFCD00190790
Molecular Formula: C10H17NO6
Molecular Weight: 247.25
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 108
Specific rotation [a]20/D: 14 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-D-glutamic Acid, Boc-D-Glu-OH

Formula: C₁₀H₁₇NO₆
MW: 247.25 g/mol
Melting Pt: 108 °C
MDL Number: MFCD00190790
CAS Number: 34404-28-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53869723e6826e8644d378db61ac3de0e875da7a&vid=53869723e6826e8644d378db61ac3de0e875da7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR