4-Butylcyclohexanecarboxylic acid (cis and trans mixture) ≥98.0%

Supplier: TCI America

Synonyms: 4-Butylcyclohexanecarboxylic acid

B2986-5G B2986-25G
TCB2986-5GEA 284.4 CAD
TCB2986-5G TCB2986-25G
4-Butylcyclohexanecarboxylic acid (cis and trans mixture) ≥98.0%
4-Butylcyclohexanecarboxylic acid (cis and trans mixture)

CAS Number: 71101-89-8
MDL Number: MFCD00460757
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 0.98

Formula: C₁₁H₂₀O₂
Density: 0.98 (20 °C)
MDL Number: MFCD00460757
CAS Number: 71101-89-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0de31b16517a6640ca1cf52c05ceaf86070ef0c2&vid=0de31b16517a6640ca1cf52c05ceaf86070ef0c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR