3-Bromo-4-fluorophenylacetic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B2976-5G B2976-25G
TCB2976-5GEA 80.58 CAD
TCB2976-5G TCB2976-25G
3-Bromo-4-fluorophenylacetic acid ≥97.0% (by GC, titration analysis)
3-Bromo-4-fluorophenylacetic acid

CAS Number: 194019-11-9
MDL Number: MFCD04038373
Molecular Formula: C8H6BrFO2
Molecular Weight: 233.04
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 67

Formula: C₈H₆BrFO₂
MW: 233.04 g/mol
Melting Pt: 67 °C
MDL Number: MFCD04038373
CAS Number: 194019-11-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11549661883db3793979c907a60495e5b23933a0&vid=11549661883db3793979c907a60495e5b23933a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR