3-Bromo-2-fluorotoluene ≥98.0%

Supplier: TCI America

B2969-5G B2969-1G
TCB2969-5GEA 121.66 CAD
TCB2969-5G TCB2969-1G
3-Bromo-2-fluorotoluene ≥98.0%
3-Bromo-2-fluorotoluene

CAS Number: 59907-12-9
MDL Number: MFCD04972694
Molecular Formula: C7H6BrF
Molecular Weight: 189.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 186
Specific Gravity (20/20): 1.52

Formula: C₇H₆BrF
Boiling Pt: 186 °C
Density: 1.52 (20 °C)
MDL Number: MFCD04972694
CAS Number: 59907-12-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d019af9d94753379fddce6554ed5bcc7a421e4e&vid=6d019af9d94753379fddce6554ed5bcc7a421e4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR