6-Benzylamino-1-hexanol ≥97.0%

Supplier: TCI America

B2968-25G B2968-5G
TCB2968-25GEA 229.1 CAD
TCB2968-25G TCB2968-5G
6-Benzylamino-1-hexanol ≥97.0%
6-Benzylamino-1-hexanol

CAS Number: 133437-08-8
Molecular Formula: C13H21NO
Molecular Weight: 207.32
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: N-(6-Hydroxyhexyl)benzylamine

Formula: C₁₃H₂₁NO
Density: 1.00 (20 °C)
CAS Number: 133437-08-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec5d60ce86b1c207d1b17dd8bb1866faead6bc6f&vid=ec5d60ce86b1c207d1b17dd8bb1866faead6bc6f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR