Nɑ-(tert-Butoxycarbonyl)-D-asparagine ≥98.0%
Supplier: TCI America
Synonyms:
Boc-D-Asn-OH, Nɑ-Boc-D-asparagine
B2964-5G
B2964-25G
TCB2964-5GEA
42.98
CAD
TCB2964-5G
TCB2964-25G
Nɑ-(tert-Butoxycarbonyl)-D-asparagine ≥98.0%
Nalpha-(tert-Butoxycarbonyl)-D-asparagine
CAS Number: 75647-01-7
MDL Number: MFCD00065558
Molecular Formula: C9H16N2O5
Molecular Weight: 232.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 7 deg (C=1, DMF)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Boc-D-asparagine, Boc-D-Asn-OH
Formula:
C₉H₁₆N₂O₅ MW: 232.24 g/mol |
MDL Number:
MFCD00065558 CAS Number: 75647-01-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...