N-(tert-Butoxycarbonyl)-D-tyrosine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B2963-5G B2963-1G
TCB2963-5GEA 413.96 CAD
TCB2963-5G TCB2963-1G
N-(tert-Butoxycarbonyl)-D-tyrosine ≥98.0% (by titrimetric analysis)
N-(tert-Butoxycarbonyl)-D-tyrosine

CAS Number: 70642-86-3
MDL Number: MFCD00063030
Molecular Formula: C14H19NO5
Molecular Weight: 281.31
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -2 deg (C=2, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-D-tyrosine, Boc-D-Tyr-OH

Formula: C₁₄H₁₉NO₅
MDL Number: MFCD00063030
CAS Number: 70642-86-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34a0c243305bdea90a501c4623c82d338ef389be&vid=34a0c243305bdea90a501c4623c82d338ef389be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR