1-Bromo-2-fluoro-5-nitrobenzene ≥95.0%

Supplier: TCI America

B2958-25G B2958-5G
TCB2958-25GEA 85.95 CAD
TCB2958-25G TCB2958-5G
1-Bromo-2-fluoro-5-nitrobenzene ≥95.0%
1-Bromo-2-fluoro-5-nitrobenzene

CAS Number: 701-45-1
MDL Number: MFCD00055433
Molecular Formula: C6H3BrFNO2
Molecular Weight: 220.00
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 57


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Bromo-4-fluoronitrobenzene

Formula: C₆H₃BrFNO₂
Melting Pt: 57 °C
MDL Number: MFCD00055433
CAS Number: 701-45-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efcec1049954fc48b8a2af26889c0d0595e999f3&vid=efcec1049954fc48b8a2af26889c0d0595e999f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR