2-Benzyloxybenzyl alcohol ≥97.0%

Supplier: TCI America

B2943-25G B2943-5G
TCB2943-25GEA 682.72 CAD
TCB2943-25G TCB2943-5G
2-Benzyloxybenzyl alcohol ≥97.0%
2-Benzyloxybenzyl alcohol

CAS Number: 3381-87-1
Molecular Formula: C14H14O2
Molecular Weight: 214.26
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 155
Freezing point (°C): 37

Formula: C₁₄H₁₄O₂
Boiling Pt: 155 °C
MDL Number: MFCD01664428
CAS Number: 3381-87-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b32fb7c2bb1ddb8f4b330034cda4ffea76a64219&vid=b32fb7c2bb1ddb8f4b330034cda4ffea76a64219&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR