Glycerol dimethacrylate(mixture of 1,2- and 1,3-form) ≥90.0% stabilized

Supplier: TCI America

Synonyms: Bis(methacryloyloxy)propanol

B2938-500G B2938-25G
TCB2938-500GEA 236.27 CAD
TCB2938-500G TCB2938-25G
Glycerol dimethacrylate(mixture of 1,2- and 1,3-form) ≥90.0% stabilized
Glycerol dimethacrylate(mixture of 1,2- and 1,3-form)

(stabilized with MEHQ)
CAS Number: 1830-78-0
MDL Number: MFCD00066299
Molecular Formula: C11H16O5
Molecular Weight: 228.24
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 172
Specific Gravity (20/20): 1.12


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Bis(methacryloyloxy)propanol

Formula: C₁₁H₁₆O₅
MW: 228.24 g/mol
Boiling Pt: 104 °C
Density: 1.12 (20 °C)
Flash Pt: 172 °C
MDL Number: MFCD00066299
CAS Number: 1830-78-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=292defbe5dfa3a06f6f18545af6962468d1b75ad&vid=292defbe5dfa3a06f6f18545af6962468d1b75ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR