1,10-Bis(acryloyloxy)decane ≥90.0% stabilised

Supplier: TCI America

Synonyms: Decamethylene Glycol Diacrylate, 1,10-Decanediol Diacrylate, Decane-1,10-diyl diacrylate

B2937-500G B2937-25G B2937-100G
TCB2937-500GEA 720.48 CAD
TCB2937-500G TCB2937-25G TCB2937-100G
1,10-Bis(acryloyloxy)decane ≥90.0% stabilised
1,10-Bis(acryloyloxy)decane

(stabilized with MEHQ)
CAS Number: 13048-34-5
MDL Number: MFCD00080453
Molecular Formula: C16H26O4
Molecular Weight: 282.38
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 181
Flash Point (°C): 200
Specific Gravity (20/20): 0.98


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,10-Decanediol Diacrylate, Decamethylene Glycol Diacrylate

Formula: C₁₆H₂₆O₄
MW: 282.38 g/mol
Boiling Pt: 181 °C
Density: 0.98 (20 °C)
Flash Pt: 200 °C
MDL Number: MFCD00080453
CAS Number: 13048-34-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2c9189d03ffcab2f2bd79455824351f633757ef&vid=e2c9189d03ffcab2f2bd79455824351f633757ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR