Bis(hexylene glycolato)diboron ≥98.0%

Supplier: TCI America

Synonyms: 4,4,4',4',6,6'-Hexamethyl-2,2'-bi-1,3,2-dioxaborinane, 4,4,4',4',6,6'-Hexamethyl-2,2'-bi(1,3,2-dioxaborinane)

B2906-1G B2906-5G
TCB2906-1GEA 135.56 CAD
TCB2906-1G TCB2906-5G
Bis(hexylene glycolato)diboron ≥98.0%
Bis(hexylene glycolato)diboron

CAS Number: 230299-21-5
MDL Number: MFCD03490499
Molecular Formula: C12H24B2O4
Molecular Weight: 253.94
Purity/Analysis Method: >98.0% (GC)
Form: Crystalline Powder
Melting point (°C): 105


Product Categories: Synthetic Organic Chemistry, Suzuki-Miyaura Cross-coupling Reaction, Boration
Other Product Names / Synonyms: 4,4,4',4',6,6'-Hexamethyl-2,2'-bi-1,3,2-dioxaborinane

Formula: C₁₂H₂₄B₂O₄
MW: 253.94 g/mol
Melting Pt: 105 °C
MDL Number: MFCD06246008
CAS Number: 230299-21-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e240d12215f600cda877de5ee2fa1eb18223f7a9&vid=e240d12215f600cda877de5ee2fa1eb18223f7a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR