N-tert-Butyl-3,5-dimethylaniline ≥98.0%

Supplier: TCI America

B2903-1G B2903-5G
TCB2903-1GEA 139.88 CAD
TCB2903-1G TCB2903-5G
N-tert-Butyl-3,5-dimethylaniline ≥98.0%
N-tert-Butyl-3,5-dimethylaniline

CAS Number: 110993-40-3
Molecular Formula: C12H19N
Molecular Weight: 177.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 70


Other Product Names / Synonyms: N-tert-Butyl-3,5-xylidine

Formula: C₁₂H₁₉N
Boiling Pt: 70 °C
MDL Number: MFCD07440135
CAS Number: 110993-40-3

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR