(R)-4-Benzyloxy-1,2-butanediol ≥97.0%

Supplier: TCI America

B2899-1G B2899-200MG
TCB2899-1GEA 456.62 CAD
TCB2899-1G TCB2899-200MG
(R)-4-Benzyloxy-1,2-butanediol ≥97.0%
(R)-4-Benzyloxy-1,2-butanediol

CAS Number: 86990-91-2
Molecular Formula: C11H16O3
Molecular Weight: 196.25
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 168
Specific rotation [a]20/D: 23 deg (C=5, EtOH)

Formula: C₁₁H₁₆O₃
Boiling Pt: 168 °C
MDL Number: MFCD10000949
CAS Number: 86990-91-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b3eccaaa43218ad34579b53c2eaf0004e51a831&vid=1b3eccaaa43218ad34579b53c2eaf0004e51a831&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR