4-tert-Butyl-1-chlorobenzene ≥98.0%

Supplier: TCI America

B2896-25G B2896-5G
TCB2896-25GEA 137.46 CAD
TCB2896-25G TCB2896-5G
4-tert-Butyl-1-chlorobenzene ≥98.0%
4-tert-Butyl-1-chlorobenzene

CAS Number: 3972-56-3
Molecular Formula: C10H13Cl
Molecular Weight: 168.66
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 216
Freezing point (°C): 23

Formula: C₁₀H₁₃Cl
Boiling Pt: 216 °C
MDL Number: MFCD00236378
CAS Number: 3972-56-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=586dcb7950153809d4fc2ee79fc405bce252cbee&vid=586dcb7950153809d4fc2ee79fc405bce252cbee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR