N-(tert-Butoxycarbonyl)-5-aminovaleric acid ≥98.0%

Supplier: TCI America

Synonyms: Boc-5-Ava-OH, BOC-5-Aminopentanoic acid, 5-(Boc-amino)valeric Acid, N-(tert-Butoxycarbonyl)-5-aminopentanoic Acid, 5-(Boc-amino)pentanoic Acid, N-Boc-5-aminovaleric Acid, 5-[(tert-Butoxycarbonyl)amino]valeric Acid, Boc-NH-C4-acid, N-Boc-5-aminopentanoic Acid, 5-[(tert-Butoxycarbonyl)amino]pentanoic Acid

B2895-25G B2895-5G
TCB2895-25GEA 477.16 CAD
TCB2895-25G TCB2895-5G
N-(tert-Butoxycarbonyl)-5-aminovaleric acid ≥98.0%
N-(tert-Butoxycarbonyl)-5-aminovaleric acid

CAS Number: 27219-07-4
MDL Number: MFCD00076903
Molecular Formula: C10H19NO4
Molecular Weight: 217.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 168
Melting point (°C): 52
Flash Point (°C): 113

Formula: C₁₀H₁₉NO₄
MW: 217.27 g/mol
Boiling Pt: 168 °C
Melting Pt: 52 °C
Flash Pt: 113 °C
MDL Number: MFCD00076903
CAS Number: 27219-07-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df749b5a02884248d57fe09060cfccf7d99fa453&vid=df749b5a02884248d57fe09060cfccf7d99fa453&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR