4-(tert-Butoxycarbonylamino)-1-butanol ≥96.0%

Supplier: TCI America

B2868-25G B2868-5G
TCB2868-25GEA 1034.9 CAD
TCB2868-25G TCB2868-5G
4-(tert-Butoxycarbonylamino)-1-butanol ≥96.0%
4-(tert-Butoxycarbonylamino)-1-butanol

CAS Number: 75178-87-9
Molecular Formula: C9H19NO3
Molecular Weight: 189.26
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Freezing point (°C): 29


Other Product Names / Synonyms: N-Boc-4-aminobutanol, tert-Butyl N-(4-Hydroxybutyl)carbamate, N-(4-Hydroxybutyl)carbamic Acid tert-Butyl Ester

Formula: C₉H₁₉NO₃
MDL Number: MFCD01861334
CAS Number: 75178-87-9

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR