Ethyl-N-acetylglycinate(Ac-Gly-OEt) ≥98.0% for synthesis

Supplier: TCI America

Synonyms: N-Acetylglycine ethyl ester, Ethyl 2-(acetylamino)acetate, Ac-Gly-OEt, Ethyl acetamidoacetate, Ethyl N-acetylglycinate

A0094-25G A0094-1G
TCA0094-025GEA 94.8 CAD
TCA0094-025G TCA0094-001G
Ethyl-N-acetylglycinate(Ac-Gly-OEt) ≥98.0% for synthesis
Ethyl N-acetylglycinate (Ac-Gly-OEt)

CAS Number: 1906-82-7
MDL Number: MFCD00009173
Molecular Formula: C6H11NO3
Molecular Weight: 145.16
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Boiling point (°C): 143
Melting point (°C): 47


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: Ac-Gly-OEt

Formula: C₆H₁₁NO₃
MW: 145.1 g/mol
Boiling Pt: 143 °C
Melting Pt: 47 °C
MDL Number: MFCD00009173
CAS Number: 1906-82-7
EINECS: 217-608-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78f459b4a0f8ef3a266892347b713cf83d4c0f35&vid=78f459b4a0f8ef3a266892347b713cf83d4c0f35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR