5-(tert-Butyldiphenylsilyloxy)-1,3-benzenedimethanol ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: [3,5-Bis(hydroxymethyl)phenoxy]-tert-butyldiphenylsilane

B2866-5G B2866-1G
TCB2866-5GEA 2575.4 CAD
TCB2866-5G TCB2866-1G
5-(tert-Butyldiphenylsilyloxy)-1,3-benzenedimethanol ≥98.0% (by HPLC)
5-(tert-Butyldiphenylsilyloxy)-1,3-benzenedimethanol

CAS Number: 482627-84-9
Molecular Formula: C24H28O3Si
Molecular Weight: 392.57
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: [3,5-Bis(hydroxymethyl)phenoxy]-tert-butyldiphenylsilane

Formula: C₂₄H₂₈O₃Si
MW: 392.57 g/mol
CAS Number: 482627-84-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d432455f09e9575bc715a1f6d1acd59bed1a02e&vid=1d432455f09e9575bc715a1f6d1acd59bed1a02e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR