9-Benzoylcarbazole ≥98.0%

Supplier: TCI America

B2864-1G B2864-5G
TCB2864-1GEA 48.98 CAD
TCB2864-1G TCB2864-5G
9-Benzoylcarbazole ≥98.0%
9-Benzoylcarbazole

CAS Number: 19264-68-7
Molecular Formula: C19H13NO
Molecular Weight: 271.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 101

Formula: C₁₉H₁₃NO
Melting Pt: 101 °C
MDL Number: MFCD00225468
CAS Number: 19264-68-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b818c19e095542ab7a836f26f766124941a7a5ff&vid=b818c19e095542ab7a836f26f766124941a7a5ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR