1,9-Bis(acryloyloxy)nonane ≥92.0% stabilized
Supplier: TCI America
B2863-500G
B2863-25G
TCB2863-500GEA
303.36
CAD
TCB2863-500G
TCB2863-25G
1,9-Bis(acryloyloxy)nonane ≥92.0% stabilized
1,9-Bis(acryloyloxy)nonane
(stabilized with MEHQ)
CAS Number: 107481-28-7
Molecular Formula: C15H24O4
Molecular Weight: 268.35
Purity/Analysis Method: >92.0% (GC)
Form: Clear Liquid
Flash Point (°C): 164
Specific Gravity (20/20): 0.99
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Nonamethylene Glycol Diacrylate, Nonanediol Diacrylate
Formula:
C₁₅H₂₄O₄ Density: 0.99 (20 °C) Flash Pt: 164 °C |
MDL Number:
MFCD00080592 CAS Number: 107481-28-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...