5-Bromo-2-thiopheneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
B2862-1G
B2862-5G
TCB2862-1GEA
116.92
CAD
TCB2862-1G
TCB2862-5G
5-Bromo-2-thiopheneboronic acid (contains varying amounts of Anhydride)
5-Bromo-2-thiopheneboronic acid
CAS Number: 162607-17-2
Molecular Formula: C4H4BBrO2S
Molecular Weight: 206.85
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 135
Storage Temperature: 0-10°C
Formula:
C₄H₄BBrO₂S Melting Pt: 135 °C |
MDL Number:
MFCD02094028 CAS Number: 162607-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...