Bis(3-amino-4-hydroxyphenyl)sulfone ≥98.0%

Supplier: TCI America

Synonyms: 3,3'-Diamino-4,4'-dihydroxydiphenyl sulphone, 3,3'-Diamino-4,4'-dihydroxydiphenyl Sulphone, 3,3'-Diamino-4,4'-dihydroxydiphenyl sulfone

B2859-5G B2859-25G
TCB2859-5GEA 325.48 CAD
TCB2859-5G TCB2859-25G
Bis(3-amino-4-hydroxyphenyl)sulfone ≥98.0%
Bis(3-amino-4-hydroxyphenyl)sulfone

CAS Number: 7545-50-8
Molecular Formula: C12H12N2O4S
Molecular Weight: 280.30
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 231


Product Categories: Materials Chemistry, Monomers, Monomers, Polyimide Raw Materials, Diphenyl Sulfones (for High-Performance Polymer Research)
Other Product Names / Synonyms: 3,3'-Diamino-4,4'-dihydroxydiphenyl Sulfone

Formula: C₁₂H₁₂N₂O₄S
MW: 280.30 g/mol
Melting Pt: 231 °C
MDL Number: MFCD00025181
CAS Number: 7545-50-8

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR