tert-Butyl [bis(4-methoxyphenyl)phosphinyloxy]carbamate ≥95.0% (by HPLC, total nitrogen)

Supplier: TCI America

B2857-1G B2857-5G
TCB2857-1GEA 163.69 CAD
TCB2857-1G TCB2857-5G
tert-Butyl [bis(4-methoxyphenyl)phosphinyloxy]carbamate ≥95.0% (by HPLC, total nitrogen)
tert-Butyl [bis(4-methoxyphenyl)phosphinyloxy]carbamate

CAS Number: 619333-95-8
Molecular Formula: C19H24NO6P
Molecular Weight: 393.38
Purity/Analysis Method: >95.0% (HPLC,N)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Amination
Other Product Names / Synonyms: [Bis(4-methoxyphenyl)phosphinyloxy]carbamic Acid tert-Butyl Ester, O-[Bis(4-methoxyphenyl)phosphinyl]-N-Boc-hydroxylamine, O-[Bis(4-methoxyphenyl)phosphinyl]-N-(tert-butoxycarbonyl)hydroxylamine

Formula: C₁₉H₂₄NO₆P
CAS Number: 619333-95-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99b6a2847233e75f09d15b035e01f15a6f69975a&vid=99b6a2847233e75f09d15b035e01f15a6f69975a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR