Disodium adipate 98,0

Supplier: TCI America

Synonyms: Disodium Adipate, Sodium adipate, Adipic Acid Disodium Salt

A0592-25G A0592-500G
TCA0592-025GEA 124.03 CAD
TCA0592-025G TCA0592-500G
Disodium adipate 98,0
Disodium adipate

CAS Number: 7486-38-6
MDL Number: MFCD00070498
Molecular Formula: C6H10O4
Molecular Weight: 190.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Other Product Names / Synonyms: Adipic Acid Disodium Salt

Formula: C₆H₁₀O₄
MW: 190.11 g/mol
MDL Number: MFCD00070498
CAS Number: 7486-38-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82fe1c9ac8ebe0c1dfd1d631ce150758ca6bb84f&vid=82fe1c9ac8ebe0c1dfd1d631ce150758ca6bb84f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR