4,4'-Biphenyldicarboxaldehyde ≥98.0%
Supplier: TCI America
Synonyms:
[1,1'-Biphenyl]-4,4'-dicarbaldehyde, 4,4'-Diformylbiphenyl
B2854-1G
B2854-5G
TCB2854-1GEA
150.1
CAD
TCB2854-1G
TCB2854-5G
4,4'-Biphenyldicarboxaldehyde ≥98.0%
4,4'-Biphenyldicarboxaldehyde
CAS Number: 66-98-8
Molecular Formula: C14H10O2
Molecular Weight: 210.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 146
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Diformylbiphenyl
Formula:
C₁₄H₁₀O₂ Melting Pt: 146 °C |
MDL Number:
MFCD00016714 CAS Number: 66-98-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...