2-(2-Bromoethoxy)tetrahydropyran ≥95.0% stabilized

Supplier: TCI America

B2850-25G B2850-5G
TCB2850-25GEA 598.82 CAD
TCB2850-25G TCB2850-5G
2-(2-Bromoethoxy)tetrahydropyran ≥95.0% stabilized
2-(2-Bromoethoxy)tetrahydropyran

(stabilized with K2CO3)
CAS Number: 17739-45-6
Molecular Formula: C7H13BrO2
Molecular Weight: 209.08
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 75
Flash Point (°C): 87
Specific Gravity (20/20): 1.38
Storage Temperature: 0-10°C

Formula: C₇H₁₃BrO₂
Boiling Pt: 75 °C
Density: 1.38 (20 °C)
Flash Pt: 87 °C
MDL Number: MFCD01321310
CAS Number: 17739-45-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=521018c8db97a4a6e70c3165a9706c910baa402d&vid=521018c8db97a4a6e70c3165a9706c910baa402d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR