10-Bromodec-1-ene ≥95.0%
Supplier: TCI America
B2849-1G
B2849-5G
TCB2849-1GEA
30.02
CAD
TCB2849-1G
TCB2849-5G
10-Bromodec-1-ene ≥95.0%
10-Bromodec-1-ene
CAS Number: 62871-09-4
Molecular Formula: C10H19Br
Molecular Weight: 219.17
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 96
Specific Gravity (20/20): 1.09
Other Product Names / Synonyms: 9-Decenyl Bromide
Formula:
C₁₀H₁₉Br Boiling Pt: 150 °C Density: 1.09 (20 °C) Flash Pt: 96 °C |
MDL Number:
MFCD00078660 CAS Number: 62871-09-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...