4-Bromo-4'-(diphenylamino)biphenyl ≥93.0%

Supplier: TCI America

Synonyms: 4'-Bromo-N,N-diphenyl-[1,1'-biphenyl]-4-amine, (4'-Bromobiphenyl-4-yl)diphenylamine

B2844-1G B2844-250MG
TCB2844-1GEA 696.78 CAD
TCB2844-1G TCB2844-250MG
4-Bromo-4'-(diphenylamino)biphenyl ≥93.0%
4-Bromo-4'-(diphenylamino)biphenyl

CAS Number: 202831-65-0
Molecular Formula: C24H18BrN
Molecular Weight: 400.32
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Melting point (°C): 129


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: (4'-Bromobiphenyl-4-yl)diphenylamine

Formula: C₂₄H₁₈BrN
Melting Pt: 129 °C
MDL Number: MFCD09038499
CAS Number: 202831-65-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=619bad6e78885265e0b31a20a0e0bfafc84306b5&vid=619bad6e78885265e0b31a20a0e0bfafc84306b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR