Betulinic acid ≥97.0%

Supplier: TCI America

Synonyms: (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-Hydroxy-5a,5b,8,8,11a-pentamethyl-1-(prop-1-en-2-yl)icosahydro-1H-cyclopenta[a]chrysene-3a-carboxylic acid, 3beta-Hydroxy-20(29)-lupaene-28-oic Acid

B2836-100MG B2836-1G
TCB2836-100MGEA 223.48 CAD
TCB2836-100MG TCB2836-1G
Betulinic acid ≥97.0%
Betulinic acid

CAS Number: 472-15-1
Molecular Formula: C30H48O3
Molecular Weight: 456.71
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 294
Specific rotation [a]20/D: 7.8 deg (C=0.9, Pyridine)


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Regenerative Medicine Research Reagents, Terpenoids (Phytochemicals), Triterpenes
Other Product Names / Synonyms: 3beta-Hydroxy-20(29)-lupaene-28-oic Acid

Formula: C₃₀H₄₈O₃
Melting Pt: 294 °C
MDL Number: MFCD00009619
CAS Number: 472-15-1

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR