Brassinazole 93,0

Supplier: TCI America

Synonyms: 1-[2-(4-Chlorophenyl)-1-(1-hydroxy-1-phenylethyl)ethyl]-1,2,4-triazole, 3-(4-Chlorophenyl)-1-methyl-1-phenyl-2-(1,2,4-triazol-1-yl)-1-propanol, 4-(4-Chlorophenyl)-2-phenyl-3-(1H-1,2,4-triazol-1-yl)butan-2-ol

B2829-100MG B2829-10MG
TCB2829-100MGEA 233.05 CAD
TCB2829-100MG TCB2829-10MG
Brassinazole 93,0
Brassinazole

CAS Number: 224047-41-0
Molecular Formula: C18H18ClN3O
Molecular Weight: 327.81
Purity/Analysis Method: >93.0% (HPLC)
Form: Crystal


Product Categories: Bioscience, Other Plant Hormones
Other Product Names / Synonyms: 1-[2-(4-Chlorophenyl)-1-(1-hydroxy-1-phenylethyl)ethyl]-1,2,4-triazole, 3-(4-Chlorophenyl)-1-methyl-1-phenyl-2-(1,2,4-triazol-1-yl)-1-propanol

Formula: C₁₈H₁₈ClN₃O
MW: 327.81 g/mol
MDL Number: MFCD08276319
CAS Number: 224047-41-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76448dfcb18fd34724935a93ed4b3042b5ca44a1&vid=76448dfcb18fd34724935a93ed4b3042b5ca44a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR