2-(2-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0%

Supplier: TCI America

Synonyms: 2-Bromophenylboronic Acid Pinacol Ester, 1-Bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

B2828-1G B2828-5G
TCB2828-1GEA 312.84 CAD
TCB2828-1G TCB2828-5G
2-(2-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0%
2-(2-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

CAS Number: 269410-06-2
Molecular Formula: C12H16BBrO2
Molecular Weight: 282.97
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 1-Bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 2-Bromophenylboronic Acid Pinacol Ester

Formula: C₁₂H₁₆BBrO₂
MDL Number: MFCD08276318
CAS Number: 269410-06-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=613734cd7819517507ea9c8caa3d5d1ad0b50258&vid=613734cd7819517507ea9c8caa3d5d1ad0b50258&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR