Benzyl-6-bromohexyl ether ≥97.0%

Supplier: TCI America

Synonyms: (((6-Bromohexyl)oxy)methyl)benzene, Benzyl 6-Bromohexyl Ether, Benzyloxyhexyl Bromide, (6-Bromohexyloxymethyl)benzene

B2820-5G B2820-25G
TCB2820-5GEA 254.54 CAD
TCB2820-5G TCB2820-25G
Benzyl-6-bromohexyl ether ≥97.0%
Benzyl-6-bromohexyl ether

CAS Number: 54247-27-7
Molecular Formula: C13H19BrO
Molecular Weight: 271.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.21


Other Product Names / Synonyms: Benzyloxyhexyl Bromide, (6-Bromohexyloxymethyl)benzene

Formula: C₁₃H₁₉BrO
Density: 1.21 (20 °C)
MDL Number: MFCD02258455
CAS Number: 54247-27-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3573aeb1e35b7286d690e7ca9a5a5bf6bdf544f6&vid=3573aeb1e35b7286d690e7ca9a5a5bf6bdf544f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR