((3-Bromopropoxy)methyl)benzene ≥95.0%

Supplier: TCI America

Synonyms: Benzyl 3-bromopropyl ether, 3-bromopropylbenzyl ether

B2818-25G B2818-5G
TCB2818-25GEA 564.06 CAD
TCB2818-25G TCB2818-5G
((3-Bromopropoxy)methyl)benzene ≥95.0%
((3-Bromopropoxy)methyl)benzene

CAS Number: 54314-84-0
MDL Number: MFCD00134570
Molecular Formula: C10H13BrO
Molecular Weight: 229.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 141
Flash Point (°C): 113
Specific Gravity (20/20): 1.31


Other Product Names / Synonyms: (3-Bromopropoxymethyl)benzene, 3-(Benzyloxy)propyl Bromide

Formula: C₁₀H₁₃BrO
MW: 229.117 g/mol
Boiling Pt: 141 °C
Density: 1.31 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00134570
CAS Number: 54314-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3eb57a5210eaaea3a563bd40bdd666499f8e1386&vid=3eb57a5210eaaea3a563bd40bdd666499f8e1386&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR