Benzyl-2-bromoethyl ether ≥97.0%

Supplier: TCI America

Synonyms: ((2-Bromoethoxy)methyl)benzene, Benzyloxyethyl Bromide, (2-Bromoethoxymethyl)benzene

B2817-5G B2817-25G
TCB2817-5GEA 55.3 CAD
TCB2817-5G TCB2817-25G
Benzyl-2-bromoethyl ether ≥97.0%
Benzyl-2-bromoethyl ether

CAS Number: 1462-37-9
MDL Number: MFCD01321307
Molecular Formula: C9H11BrO
Molecular Weight: 215.09
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 113
Specific Gravity (20/20): 1.36


Other Product Names / Synonyms: (2-Bromoethoxymethyl)benzene, Benzyloxyethyl Bromide

Formula: C₉H₁₁BrO
Boiling Pt: 95 °C
Density: 1.36 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD01321307
CAS Number: 1462-37-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f44006f31f4ad9015ab1498d3f7c549a777b0ab7&vid=f44006f31f4ad9015ab1498d3f7c549a777b0ab7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR