18-Bromo-1-octadecene ≥93.0%
Supplier: TCI America
B2816-1G
B2816-5G
TCB2816-1GEA
368.14
CAD
TCB2816-1G
TCB2816-5G
18-Bromo-1-octadecene ≥93.0%
18-Bromo-1-octadecene
CAS Number: 172656-11-0
Molecular Formula: C18H35Br
Molecular Weight: 331.38
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.00
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 17-Octadecenyl Bromide
Formula:
C₁₈H₃₅Br Density: 1.00 (20 °C) |
CAS Number:
172656-11-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...