Benzyl dimethylphosphonoacetate ≥97.0%
Supplier: TCI America
Synonyms:
Dimethylphosphonoacetic Acid Benzyl Ester, Dimethyl (Cbz-methyl)phosphonate, Benzyl 2-(dimethoxyphosphoryl)acetate, Dimethyl (Benzyloxycarbonylmethyl)phosphonate
CAS Number: 57443-18-2
Molecular Formula: C11H15O5P
Molecular Weight: 258.21
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.23
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Dimethyl (Benzyloxycarbonylmethyl)phosphonate, Dimethyl (Cbz-methyl)phosphonate, Dimethylphosphonoacetic Acid Benzyl Ester
Formula:
C₁₁H₁₅O₅P Density: 1.23 (20 °C) |
MDL Number:
MFCD01872507 CAS Number: 57443-18-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...