Azo Blue
Supplier: TCI America
Synonyms:
Direct Violet 39
A0587-500G
A0587-25G
TCA0587-500GEA
520.61
CAD
TCA0587-500G
TCA0587-025G
Azo Blue
Azo Blue
CAS Number: 6059-34-3
MDL Number: MFCD00059133
Molecular Formula: C34H26N4O8S2
Molecular Weight: 726.69
Form: Crystal
Color: Black
Other Product Names / Synonyms: Direct Violet 39
Formula:
C₃₄H₂₆N₄O₈S₂ MW: 726.69 g/mol |
MDL Number:
MFCD00059133 CAS Number: 6059-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...