N-(tert-Butoxycarbonyl)-2-methyl-1,3-diaminopropane ≥98.0%

Supplier: TCI America

Synonyms: N-(3-Amino-2-methylpropyl)carbamic Acid tert-Butyl Ester, N-Boc-2-methyl-1,3-diaminopropane, N-Boc-2-methyl-1,3-propanediamine, tert-Butyl N-(3-Amino-2-methylpropyl)carbamate, N-(tert-Butoxycarbonyl)-2-methyl-1,3-propanediamine

B2810-1G B2810-5G
TCB2810-1GEA 185.18 CAD
TCB2810-1G TCB2810-5G
N-(tert-Butoxycarbonyl)-2-methyl-1,3-diaminopropane ≥98.0%
N-(tert-Butoxycarbonyl)-2-methyl-1,3-diaminopropane

CAS Number: 480452-05-9
Molecular Formula: C9H20N2O2
Molecular Weight: 188.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 0.98


Product Categories: Bioscience, Chemical Building Blocks for Polyamines

Formula: C₉H₂₀N₂O₂
MW: 188.27 g/mol
Density: 0.98 (20 °C)
MDL Number: MFCD06804536
CAS Number: 480452-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82db8ac952fda9ae55179c078d8725153c792f79&vid=82db8ac952fda9ae55179c078d8725153c792f79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR