4-Bromopyrene ≥95.0%
Supplier: TCI America
B2807-5G
B2807-1G
TCB2807-5GEA
3016.54
CAD
TCB2807-5G
TCB2807-1G
4-Bromopyrene ≥95.0%
4-Bromopyrene
CAS Number: 1732-26-9
Molecular Formula: C16H9Br
Molecular Weight: 281.15
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 152
Formula:
C₁₆H₉Br Melting Pt: 152 °C |
MDL Number:
MFCD00029250 CAS Number: 1732-26-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...