Bis(2,2-diethoxyethyl)disulfide ≥90.0%

Supplier: TCI America

B2806-1G B2806-5G
TCB2806-1GEA 143.78 CAD
TCB2806-1G TCB2806-5G
Bis(2,2-diethoxyethyl)disulfide ≥90.0%
Bis(2,2-diethoxyethyl)disulfide

CAS Number: 76505-71-0
Molecular Formula: C12H26O4S2
Molecular Weight: 298.46
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Specific Gravity (20/20): 1.05
Storage Temperature: <0°C

Formula: C₁₂H₂₆O₄S₂
MW: 298.46 g/mol
Boiling Pt: 155 °C
Density: 1.05 (20 °C)
CAS Number: 76505-71-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0963d35feb031c601c7f7bcf18d3bb0754967b1&vid=f0963d35feb031c601c7f7bcf18d3bb0754967b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR