9-Benzylcarbazole-3-carboxaldehyde ≥97.0% (by total nitrogen basis)

Supplier: TCI America

B2804-1G B2804-5G
TCB2804-1GEA 198.29 CAD
TCB2804-1G TCB2804-5G
9-Benzylcarbazole-3-carboxaldehyde ≥97.0% (by total nitrogen basis)
9-Benzylcarbazole-3-carboxaldehyde

CAS Number: 54117-37-2
Molecular Formula: C20H15NO
Molecular Weight: 285.35
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 134


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 9-Benzyl-3-formylcarbazole

Formula: C₂₀H₁₅NO
Melting Pt: 134 °C
MDL Number: MFCD00403522
CAS Number: 54117-37-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99ab6c71b610c3cc7ff76303570a0b8aa162b526&vid=99ab6c71b610c3cc7ff76303570a0b8aa162b526&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR