Bis(4-tert-butylphenyl)amine ≥90.0%
Supplier: TCI America
B2803-25G
B2803-500G
TCB2803-25GEA
59.57
CAD
TCB2803-25G
TCB2803-500G
Bis(4-tert-butylphenyl)amine ≥90.0%
Bis(4-tert-butylphenyl)amine
CAS Number: 4627-22-9
Molecular Formula: C20H27N
Molecular Weight: 281.44
Purity/Analysis Method: >90.0% (GC)
Form: Crystal
Boiling point (°C): 195
Melting point (°C): 107
Flash Point (°C): 193
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 4,4'-Di-tert-butyldiphenylamine
Formula:
C₂₀H₂₇N Boiling Pt: 195 °C Melting Pt: 107 °C Flash Pt: 193 °C |
MDL Number:
MFCD00417391 CAS Number: 4627-22-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...