Benzyl-4-bromobutyl ether ≥96.0%

Supplier: TCI America

Synonyms: Benzyl 4-bromobutyl ether, (4-Bromobutoxymethyl)benzene, ((4-Bromobutoxy)methyl)benzene, 4-Benzyloxybutyl Bromide

B2802-25G B2802-5G
TCB2802-25GEA 856.2 CAD
TCB2802-25G TCB2802-5G
Benzyl-4-bromobutyl ether ≥96.0%
Benzyl-4-bromobutyl ether

CAS Number: 60789-54-0
Molecular Formula: C11H15BrO
Molecular Weight: 243.14
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 113
Specific Gravity (20/20): 1.28


Other Product Names / Synonyms: 4-Benzyloxybutyl Bromide, (4-Bromobutoxymethyl)benzene

Formula: C₁₁H₁₅BrO
Boiling Pt: 115 °C
Density: 1.28 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00155128
CAS Number: 60789-54-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d12a062ec7e9e2d0fa403efc5c63ea0790ac0dc&vid=7d12a062ec7e9e2d0fa403efc5c63ea0790ac0dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR