9,10-Bis(diethylphosphonomethyl)anthracene ≥98.0%

Supplier: TCI America

Synonyms: [Anthracene-9,10-diylbis(methylene)]bisphosphonic Acid Tetraethyl Ester, Tetraethyl [Anthracene-9,10-diylbis(methylene)]bisphosphonate, Tetraethyl (anthracene-9,10-diylbis(methylene))bis(phosphonate)

B2801-1G B2801-5G
TCB2801-1GEA 72.68 CAD
TCB2801-1G TCB2801-5G
9,10-Bis(diethylphosphonomethyl)anthracene ≥98.0%
9,10-Bis(diethylphosphonomethyl)anthracene

CAS Number: 60974-92-7
Molecular Formula: C24H32O6P2
Molecular Weight: 478.46
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 160


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: [Anthracene-9,10-diylbis(methylene)]bisphosphonic Acid Tetraethyl Ester, Tetraethyl [Anthracene-9,10-diylbis(methylene)]bisphosphonate

Formula: C₂₄H₃₂O₆P₂
MW: 478.46 g/mol
Melting Pt: 160 °C
MDL Number: MFCD08276315
CAS Number: 60974-92-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd2381cf61a81a2ee07fd178b6d5981b7db933b5&vid=dd2381cf61a81a2ee07fd178b6d5981b7db933b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR