5-Bromo-2-hydroxy-3-picoline ≥98.0%
Supplier: TCI America
Synonyms:
5-Bromo-3-methylpyridin-2-ol, 5-Bromo-2-hydroxy-3-methylpyridine, 5-Bromo-3-methylpyridin-2(1H)-one
B2797-1G
B2797-5G
TCB2797-1GEA
26.86
CAD
TCB2797-1G
TCB2797-5G
5-Bromo-2-hydroxy-3-picoline ≥98.0%
5-Bromo-2-hydroxy-3-picoline
CAS Number: 89488-30-2
Molecular Formula: C6H6BrNO
Molecular Weight: 188.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 172
Other Product Names / Synonyms: 5-Bromo-2-hydroxy-3-picoline, 5-Bromo-3-methyl-2-pyridinol, 5-Bromo-3-methyl-2-pyridone
Formula:
C₆H₆BrNO MW: 188.02 g/mol Melting Pt: 172 °C |
MDL Number:
MFCD03427657 CAS Number: 89488-30-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...